Cyclohexanemethylamine Hydroiodide ≥98.0% (by total nitrogen and titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Cyclohexanemethylammonium iodide, (Aminomethyl)cyclohexane Hydroiodide, Cyclohexylmethylamine Hydroiodide
C3425-1G
C3425-5G
TCIAC3425-1GEA
56.1
EUR
TCIAC3425-1G
TCIAC3425-5G
Cyclohexanemethylamine Hydroiodide ≥98.0% (by total nitrogen and titration analysis)
Cyclohexanemethylamine Hydroiodide
Formule:
C₇H₁₅N·HI MW: 241.12 g/mol |
CAS nummer:
2153504-15-3 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...