Chloro(3-chloropropyl)dimethylsilane ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

C3426-1ML C3426-5ML
TCIAC3426-1MLEA 24.2 EUR
TCIAC3426-1ML TCIAC3426-5ML
Chloro(3-chloropropyl)dimethylsilane ≥96.0% (by GC)
Chloro(3-chloropropyl)dimethylsilane
Formule: C₅H₁₂Cl₂Si
Kookpunt: 179°C
MDL Nummer: MFCD00042612
CAS nummer: 10605-40-0
EINECS: 234-234-1
UN: 2987
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=baad3c6da63408994652e13d4b9022af38b7eb5c&vid=baad3c6da63408994652e13d4b9022af38b7eb5c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR