4-Chlorobenzo[b]thiophene-2-carboxylic acid ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

C3430-1G C3430-5G
TCIAC3430-1GEA 74.8 EUR
TCIAC3430-1G TCIAC3430-5G
4-Chlorobenzo[b]thiophene-2-carboxylic acid ≥97.0% (by GC)
4-Chlorobenzo[b]thiophene-2-carboxylic acid
Formule: C₉H₅ClO₂S
MDL Nummer: MFCD06655155
CAS nummer: 23967-57-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR