N-Carbobenzoxy-1,12-diaminododecane hydrochloride ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

C3472-1G C3472-5G
TCIAC3472-1GEA 123 EUR
TCIAC3472-1G TCIAC3472-5G
N-Carbobenzoxy-1,12-diaminododecane hydrochloride ≥98.0% (by HPLC)
N-Carbobenzoxy-1,12-diaminododecane hydrochloride
Formule: C₂₀H₃₄N₂O₂·HCl
MDL Nummer: MFCD08272292
CAS nummer: 1051420-16-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98c7bfdfaf8da51ec95555414d0bcb395b9ba708&vid=98c7bfdfaf8da51ec95555414d0bcb395b9ba708&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 173.0 to 178.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR