Scutellarein ≥95.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 4',5,6,7-Tetrahydroxyflavone, 5,6,7-Trihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one

C3507-10MG C3507-50MG
TCIAC3507-10MGEA 126 EUR
TCIAC3507-10MG TCIAC3507-50MG
Scutellarein ≥95.0% (by HPLC)
Scutellarein
Formule: C₁₅H₁₀O₆
Smeltpunt: 347°C
MDL Nummer: MFCD00017692
CAS nummer: 529-53-3
Merck Index: 14,08412

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 95.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR