Cyclohexylamine Hydroiodide ≥95.0% (by total nitrogen and titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
C3532-5G
C3532-1G
TCIAC3532-5GEA
103
EUR
TCIAC3532-5G
TCIAC3532-1G
Cyclohexylamine Hydroiodide ≥95.0% (by total nitrogen and titration analysis)
Cyclohexylamine Hydroiodide
Formule:
C₆H₁₃N·HI Smeltpunt: 194°C |
CAS nummer:
45492-87-3 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 95.0 % |
Purity (Argentmetric Titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...