1-Chloor-2-propanol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 1-Chloropropan-2-ol, Propylene chlorohydrin, Chloropropanol

C3533-1G C3533-5G
TCIAC3533-1GEA 60.5 EUR
TCIAC3533-1G TCIAC3533-5G
1-Chloor-2-propanol ≥98.0% (by GC)
1-Chloor-2-propanol
Formule: C₃H₇ClO
MW: 94.54 g/mol
Kookpunt: 132°C
Vlampunt: 52°C
MDL Nummer: MFCD00004530
CAS nummer: 127-00-4
UN: 2611
ADR: 6.1 II
Merck Index: 14,07851

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR