5-ROX ≥95.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 5-ROX, 5-Carboxy-X-rhodamine

C3538-10MG C3538-50MG
TCIAC3538-10MGEA 96.8 EUR
TCIAC3538-10MG TCIAC3538-50MG
5-ROX ≥95.0% (by HPLC)
5-ROX
Formule: C₃₃H₃₀N₂O₅
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD28355897
CAS nummer: 216699-35-3

Order Now

Specification Test Results

Appearance Gray to Dark purple to Black powder to crystal
Purity (HPLC) min. 95.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR