4-Chloro-1-butene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

C3611-5G C3611-25G
TCIAC3611-5GEA 50.6 EUR
TCIAC3611-5G TCIAC3611-25G
4-Chloro-1-butene ≥98.0% (by GC)
4-Chloro-1-butene
Formule: C₄H₇Cl
Kookpunt: 75°C
MDL Nummer: MFCD00039386
CAS nummer: 927-73-1
EINECS: 213-160-3
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR