(E)-Cinnamyl Benzoate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

C3650-100G C3650-25G
TCIAC3650-100GEA 217 EUR
TCIAC3650-100G TCIAC3650-25G
(E)-Cinnamyl Benzoate ≥98.0% (by GC)
(E)-Cinnamyl Benzoate
Formule: C₁₆H₁₄O₂
Kookpunt: 335°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00475021
CAS nummer: 50555-04-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 37.0 to 41.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR