Dimethyl sulphoxide-[D₆] ≥99.0%
Leverancier: TCI EUROPE
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
TCIAD0381-25GEA
345
EUR
TCIAD0381-25G
Dimethyl sulphoxide-[D₆] ≥99.0%
Dimethyl sulphoxide-[D₆]
Formule:
D₃CS(O)CD₃ MW: 84,09 g/mol Kookpunt: 74°C Vlampunt: 95°C |
MDL Nummer:
MFCD00002090 CAS nummer: 2206-27-1 EINECS: 218-617-0 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Degree of deuteration | min. 99.9 atom%D |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...