N,N-Dimethyl-2-naphthylamine ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D0756-1G
D0756-5G
TCIAD0756-1GEA
62.7
EUR
TCIAD0756-1G
TCIAD0756-5G
N,N-Dimethyl-2-naphthylamine ≥98.0% (by GC)
N,N-Dimethyl-2-naphthylamine
Formule:
C₁₂H₁₃N Kookpunt: 305°C |
MDL Nummer:
MFCD00059550 CAS nummer: 2436-85-3 EINECS: 219-432-8 |
Specification Test Results
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Melting point | 45.0 to 47.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...