2,4-Diphenyloxazole ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

D0901-1G D0901-25G D0901-5G
TCIAD0901-1GEA 51.7 EUR
TCIAD0901-1G TCIAD0901-25G TCIAD0901-5G
2,4-Diphenyloxazole ≥99.0% (by GC)
2,4-Diphenyloxazole
Formule: C₁₅H₁₁NO
Kookpunt: 340°C
MDL Nummer: MFCD00016890
CAS nummer: 838-41-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 103.0 to 106.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR