β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) hydrate for biochemical research
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: beta-NADH disodium salt hydrate, beta-Nicotinamide adenine dinucleotide disodium salt hydrate, β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form) hydrate
D0920-1G D0920-100MG D0920-5G
TCIAD0920-1GEA 77 EUR
TCIAD0920-1G TCIAD0920-100MG TCIAD0920-5G
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) hydrate for biochemical research
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) hydrate
Formule: C₂₁H₂₇N₇Na₂O₁₄P₂·H₂O
MW: 727.43 g/mol
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00036200
CAS nummer: 606-68-8
EINECS: 210-123-3
Order Now
Specification Test Results
Appearance White to Orange to Green powder to crystal
abs-ratio (a/b) 0.79 to 0.85 (E250/E260, in pH10.0)
abs-ratio (2a/2b) 0.21 to 0.24 (E280/E260, in pH10.0)
abs-ratio (3a/3b) 0.42 to 0.44 (E340/E260, in pH10.0)
Molar extinction coefficient 14400 to 15400 (pH10.0, calcd.on anh.substance)