2-(Dimethylamino)-2-methyl-1-propanol ≥97.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
2-(Dimethylamino)isobutanol
D1098-25ML
D1098-500ML
TCIAD1098-25MLEA
24.2
EUR
TCIAD1098-25ML
TCIAD1098-500ML
2-(Dimethylamino)-2-methyl-1-propanol ≥97.0% (by GC, titration analysis)
2-(Dimethylamino)-2-methyl-1-propanol
Formule:
C₆H₁₅NO Kookpunt: 163°C Vlampunt: 53°C |
MDL Nummer:
MFCD00021978 CAS nummer: 7005-47-2 EINECS: 230-279-6 UN: 2734 ADR: 8,II |
Specification Test Results
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...