Diethyl diallylmalonate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Diethyl 2,2-diallylmalonate, Diallylmalonic Acid Diethyl Ester
D1183-25ML
D1183-100ML
TCIAD1183-25MLEA
138
EUR
TCIAD1183-25ML
TCIAD1183-100ML
Diethyl diallylmalonate ≥98.0% (by GC)
Diethyl diallylmalonate
Formule:
C₁₃H₂₀O₄ MW: 240.30 g/mol Kookpunt: 128°C |
MDL Nummer:
MFCD00009126 CAS nummer: 3195-24-2 EINECS: 221-696-4 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...