Dabsyl chloride ≥98.0% (by titrimetric analysis) N-protecting agent, for peptide research

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: DABS-Cl, 4-Dimethylaminoazobenzene-4'-sulphonyl Chloride, 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonyl chloride

D1382-1G D1382-25G
TCIAD1382-1GEA 74.8 EUR
TCIAD1382-1G TCIAD1382-25G
Dabsyl chloride ≥98.0% (by titrimetric analysis) N-protecting agent, for peptide research
Dabsyl chloride
Formule: C₁₄H₁₄ClN₃O₂S
MW: 323.80 g/mol
MDL Nummer: MFCD00007444
CAS nummer: 56512-49-3
EINECS: 260-235-1
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Appearance Orange to Brown to Dark red powder to crystaline
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR