Dansylglycine ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

D1501-1G D1501-5G D1501-100MG
TCIAD1501-1GEA 134 EUR
TCIAD1501-1G TCIAD1501-5G TCIAD1501-100MG
Dansylglycine ≥98.0% (by titrimetric analysis)
Dansylglycine
Formule: C₁₄H₁₆N₂O₄S
MDL Nummer: MFCD00037734
CAS nummer: 1091-85-6
EINECS: 214-129-7

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 156.0 to 161.0 deg-C
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR