Dimethylisopropylchlorosilane ≥90.0% (by GC) dimethyl-isopropyl-silylating agent

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Chlorodimethylisopropylsilane, Isopropyldimethylchlorosilane

D1594-25ML D1594-5ML
TCIAD1594-25MLEA 149 EUR
TCIAD1594-25ML TCIAD1594-5ML
Dimethylisopropylchlorosilane ≥90.0% (by GC) dimethyl-isopropyl-silylating agent
Dimethylisopropylchlorosilane
Formule: C₅H₁₃ClSi
Kookpunt: 112°C
Vlampunt: 15°C
MDL Nummer: MFCD00009917
CAS nummer: 3634-56-8
UN: 2985
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR