Stearyl-3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D1644-25G D1644-500G D1644-100G
TCIAD1644-25GEA 17.6 EUR
TCIAD1644-25G TCIAD1644-500G TCIAD1644-100G
Stearyl-3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate ≥98.0% (by GC)
Stearyl-3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate
Formule: C₃₅H₆₂O₃
MDL Nummer: MFCD00017498
CAS nummer: 2082-79-3
EINECS: 218-216-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 55.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR