2-[2-(Dimethylamino)ethoxy]ethanol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Ethylene Glycol Mono[2-(dimethylamino)ethyl] Ether, 2-(2-(Dimethylamino)ethoxy)ethanol

D1756-25ML D1756-500ML
TCIAD1756-25MLEA 17.6 EUR
TCIAD1756-25ML TCIAD1756-500ML
2-[2-(Dimethylamino)ethoxy]ethanol ≥98.0% (by GC)
2-[2-(Dimethylamino)ethoxy]ethanol
Formule: C₆H₁₅NO₂
MW: 133.19 g/mol
Vlampunt: 93°C
MDL Nummer: MFCD00059602
CAS nummer: 1704-62-7
EINECS: 216-940-1

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR