Nickel(II) dibutyldithiocarbamate ≥97.0% (by titrimetric analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Nickel dibutyldithiocarbamate, Nickel(II) Bis(dibutyldithiocarbamate), Dibutyldithiocarbamic Acid Nickel(II) Salt
D1781-25G D1781-500G
TCIAD1781-25GEA 17.6 EUR
TCIAD1781-25G TCIAD1781-500G
Nickel(II) dibutyldithiocarbamate ≥97.0% (by titrimetric analysis)
Nickel(II) dibutyldithiocarbamate
Formule: C₉H₁₉NS₂
MW: 467.43 g/mol
MDL Nummer: MFCD00067273
CAS nummer: 13927-77-0
EINECS: 237-696-2
Order Now
Specification Test Results
Appearance Green to Dark green powder to crystal
Purity (Chelometric Titration) min. 97.0 %