N-[3-(Dimethylamino)propyl]acrylamide ≥98.0% (by GC, titration analysis) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing


Stabilisatie: Stabilised with 4-methoxyphenol (MEHQ)

D1785-25G D1785-500G D1785-100G
TCIAD1785-25GEA 19.8 EUR
TCIAD1785-25G TCIAD1785-500G TCIAD1785-100G
N-[3-(Dimethylamino)propyl]acrylamide ≥98.0% (by GC, titration analysis) stabilised
N-[3-(Dimethylamino)propyl]acrylamide
Formule: C₈H₁₆N₂O
Kookpunt: 142°C
Vlampunt: 140°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00059902
CAS nummer: 3845-76-9
EINECS: 223-342-4

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR