4-Dimethylaminopyridinium bromide perbromide ≥97.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
4-(Dimethylamino)pyridine tribromide
D1787-25G
D1787-5G
TCIAD1787-25GEA
105
EUR
TCIAD1787-25G
TCIAD1787-5G
4-Dimethylaminopyridinium bromide perbromide ≥97.0% (by titrimetric analysis)
4-Dimethylaminopyridinium bromide perbromide
Formule:
C₇H₁₁Br₃N₂ MW: 362.89 g/mol |
MDL Nummer:
MFCD00043439 CAS nummer: 92976-81-3 UN: 3261 ADR: 8 III |
Specification Test Results
Appearance | Light yellow to Brown powder to crystal |
Purity (Iodometric Titration) | min. 97.0 % |
Melting point | 130.0 to 133.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...