1,3-Diaminoguanidine hydrochloride ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
D1912-25G D1912-5G
TCIAD1912-25GEA 56.1 EUR
TCIAD1912-25G TCIAD1912-5G
1,3-Diaminoguanidine hydrochloride ≥98.0% (by titrimetric analysis)
1,3-Diaminoguanidine hydrochloride
Formule: CH₇N₅
MDL Nummer: MFCD00012948
CAS nummer: 36062-19-8
EINECS: 252-854-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a71ba0d5bb244efaaf18b8d6dcaa0aedcc37fa6&vid=6a71ba0d5bb244efaaf18b8d6dcaa0aedcc37fa6&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water very faint turbidity