Bis(2-ethylhexyl)dodecanedioate ≥94.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

D2092-25ML D2092-500ML
TCIAD2092-25MLEA 17.6 EUR
TCIAD2092-25ML TCIAD2092-500ML
Bis(2-ethylhexyl)dodecanedioate ≥94.0% (by GC)
Bis(2-ethylhexyl)dodecanedioate
Formule: C₂₈H₅₄O₄
Kookpunt: 267°C
Vlampunt: 230°C
MDL Nummer: MFCD00144216
CAS nummer: 19074-24-9
EINECS: 242-798-5

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR