1,6-Diaminopyrene ≥98.0% (by total nitrogen basis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D2160-100MG D2160-500MG
TCIAD2160-100MGEA 260 EUR
TCIAD2160-100MG TCIAD2160-500MG
1,6-Diaminopyrene ≥98.0% (by total nitrogen basis)
1,6-Diaminopyrene
Formule: C₁₆H₁₂N₂
MDL Nummer: MFCD00142568
CAS nummer: 14923-84-3

Order Now

Specification Test Results

Appearance Green to Dark green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR