4-(2,5-Dioxotetrahydrofuran-3-yl)-1,2,3,4-tetrahydronaphthalene-1,2-dicarboxylic anhydride ≥97.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
D2192-25G
D2192-5G
TCIAD2192-25GEA
191
EUR
TCIAD2192-25G
TCIAD2192-5G
4-(2,5-Dioxotetrahydrofuran-3-yl)-1,2,3,4-tetrahydronaphthalene-1,2-dicarboxylic anhydride ≥97.0% (by HPLC, titration analysis)
4-(2,5-Dioxotetrahydrofuran-3-yl)-1,2,3,4-tetrahydronaphthalene-1,2-dicarboxylic anhydride
Formule:
C₁₆H₁₂O₆ |
MDL Nummer:
MFCD00030105 CAS nummer: 13912-65-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Methanolysis Method) | min. 97.0 % |
Melting point | 202.0 to 206.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...