Bis(2-pyridylmethyl)amine ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
alpha,alpha'-Iminodi(2-picoline), Bis(pyridin-2-ylmethyl)amine, 2,2'-Dipicolylamine
D2228-25G
D2228-5G
TCIAD2228-25GEA
245
EUR
TCIAD2228-25G
TCIAD2228-5G
Bis(2-pyridylmethyl)amine ≥98.0% (by titrimetric analysis)
Bis(2-pyridylmethyl)amine
Formule:
C₁₂H₁₃N₃ MW: 199.26 g/mol Kookpunt: 144°C |
MDL Nummer:
MFCD00129044 CAS nummer: 1539-42-0 EINECS: 216-266-8 |
Specification Test Results
Appearance | White to Yellow to Green clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...