Didecylmethylamine ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

D2311-25ML D2311-500ML
TCIAD2311-25MLEA 17.6 EUR
TCIAD2311-25ML TCIAD2311-500ML
Didecylmethylamine ≥95.0% (by GC)
Didecylmethylamine
Formule: C₂₁H₄₅N
Kookpunt: 145°C
Smeltpunt: –7°C
MDL Nummer: MFCD00077727
CAS nummer: 7396-58-9
EINECS: 230-990-1
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR