1,3-Diphenylpropanetrione ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

D2325-1G D2325-5G
TCIAD2325-1GEA 156 EUR
TCIAD2325-1G TCIAD2325-5G
1,3-Diphenylpropanetrione ≥98.0% (by GC)
1,3-Diphenylpropanetrione
Formule: C₁₅H₁₀O₃
MDL Nummer: MFCD00191426
CAS nummer: 643-75-4

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 69.0 to 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR