1,3-Dichloro-1,1,3,3-tetramethyldisiloxane ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
D2334-25G D2334-5G
TCIAD2334-25GEA 149 EUR
TCIAD2334-25G TCIAD2334-5G
1,3-Dichloro-1,1,3,3-tetramethyldisiloxane ≥97.0% (by GC)
1,3-Dichloro-1,1,3,3-tetramethyldisiloxane
Formule: C₄H₁₂Cl₂OSi₂
Kookpunt: 138°C
Vlampunt: 43°C
MDL Nummer: MFCD00018088
CAS nummer: 2401-73-2
EINECS: 219-278-1
UN: 2986
ADR: 8,II
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %