(R,R)-(-)-2,3-Dimethoxy-1,4-bis(dimethylamino)butane ≥90.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: (R,R)-(-)-DDB, (R,R)-(-)-2,3-Dimethoxy-N,N,N',N'-tetramethyl-1,4-butanediamine

D2395-1G D2395-5G
TCIAD2395-1GEA 176 EUR
TCIAD2395-1G TCIAD2395-5G
(R,R)-(-)-2,3-Dimethoxy-1,4-bis(dimethylamino)butane ≥90.0% (by GC)
(R,R)-(-)-2,3-Dimethoxy-1,4-bis(dimethylamino)butane
Formule: C₁₀H₂₄N₂O₂
MDL Nummer: MFCD00008331
CAS nummer: 26549-22-4
EINECS: 247-795-2

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 90.0 %
Optical purity (GC) min. 98.0 ee%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR