2,5-Diformylfuran ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D2408-1G
D2408-100MG
D2408-5G
TCIAD2408-1GEA
253
EUR
TCIAD2408-1G
TCIAD2408-100MG
TCIAD2408-5G
2,5-Diformylfuran ≥98.0% (by GC)
2,5-Diformylfuran
Formule:
C₆H₄O₃ Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00671517 CAS nummer: 823-82-5 EINECS: 212-520-7 |
Specification Test Results
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 109.0 to 113.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...