Dodecafluorosuberic acid ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D2465-25G D2465-5G
TCIAD2465-25GEA 230 EUR
TCIAD2465-25G TCIAD2465-5G
Dodecafluorosuberic acid ≥97.0% (by titrimetric analysis)
Dodecafluorosuberic acid
Formule: C₈H₂F₁₂O₄
MDL Nummer: MFCD00153146
CAS nummer: 678-45-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 97.0 %
Melting point 143.0 to 148.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR