2,4-Diphenyl-4-methyl-1-pentene ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 4-Methyl-2,4-diphenyl-1-pentene, (4-Methylpent-1-ene-2,4-diyl)dibenzene

D2497-25ML D2497-500ML D2497-100ML
TCIAD2497-25MLEA 19.8 EUR
TCIAD2497-25ML TCIAD2497-500ML TCIAD2497-100ML
2,4-Diphenyl-4-methyl-1-pentene ≥95.0% (by GC)
2,4-Diphenyl-4-methyl-1-pentene
Formule: C₁₈H₂₀
MW: 236.36 g/mol
Kookpunt: 175°C
Vlampunt: 162°C
MDL Nummer: MFCD00044027
CAS nummer: 6362-80-7
EINECS: 228-846-8
UN: 3082
ADR: 9 III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR