Ethoxydiphenylphosphine ≥93.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D2558-25G D2558-5G
TCIAD2558-25GEA 113 EUR
TCIAD2558-25G TCIAD2558-5G
Ethoxydiphenylphosphine ≥93.0% (by titrimetric analysis)
Ethoxydiphenylphosphine
Formule: C₁₄H₁₅OP
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00009087
CAS nummer: 719-80-2
EINECS: 211-951-8

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (Iodometric Titration) min. 93.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR