4-(4-Diethylaminophenylazo)pyridine ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D2568-1G D2568-5G
TCIAD2568-1GEA 86.9 EUR
TCIAD2568-1G TCIAD2568-5G
4-(4-Diethylaminophenylazo)pyridine ≥98.0% (by HPLC, titration analysis)
4-(4-Diethylaminophenylazo)pyridine
Formule: C₁₅H₁₈N₄
MDL Nummer: MFCD01631306
CAS nummer: 89762-42-5

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR