Dichloro(p-cymene)ruthenium(II) dimer ≥95.0% (by titrimetric analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Di(μ-chloro)dichlorobis(p-cymene)diruthenium(II)
D2751-1G D2751-5G
TCIAD2751-1GEA 75.9 EUR
TCIAD2751-1G TCIAD2751-5G
Dichloro(p-cymene)ruthenium(II) dimer ≥95.0% (by titrimetric analysis)
Dichloro(p-cymene)ruthenium(II) dimer
Formule: C₂₀H₂₈Cl₄Ru₂
MW: 612.38 g/mol
MDL Nummer: MFCD00064793
CAS nummer: 52462-29-0
EINECS: 435-530-5
Order Now
Specification Test Results
Appearance Light yellow to Brown powder to crystal
Purity (Argentmetric Titration) min. 95.0 %