3-Hydroxytyrosol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2-(3,4-Dihydroxyphenyl)ethanol, Hydroxytyrosol, DOPET, Homoprotocatechuyl alcohol, 2-(3,4-Dihydroxyphenyl)ethyl alcohol, 3,4-Dihydroxyphenethyl Alcohol

D2756-1G D2756-5G
TCIAD2756-1GEA 190 EUR
TCIAD2756-1G TCIAD2756-5G
3-Hydroxytyrosol ≥98.0% (by GC)
3-Hydroxytyrosol
Formule: C₈H₁₀O₃
MDL Nummer: MFCD01320529
CAS nummer: 10597-60-1

Order Now

Specification Test Results

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR