2,6-Dihydroxy-4-methylbenzoic acid

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D2770-25G D2770-5G
TCIAD2770-25GEA 302 EUR
TCIAD2770-25G TCIAD2770-5G
2,6-Dihydroxy-4-methylbenzoic acid
2,6-Dihydroxy-4-methylbenzoic acid
Formule: C₈H₈O₄
MDL Nummer: MFCD00184686
CAS nummer: 480-67-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR