N,N-Diethyl-2,3-dimethylbenzamide ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D2956-25G D2956-5G
TCIAD2956-25GEA 167 EUR
TCIAD2956-25G TCIAD2956-5G
N,N-Diethyl-2,3-dimethylbenzamide ≥98.0% (by GC)
N,N-Diethyl-2,3-dimethylbenzamide
Formule: C₁₃H₁₉NO
MDL Nummer: MFCD06797096
CAS nummer: 57806-76-5
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %