1,6-Dibromopyrene ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D3167-1G
D3167-5G
TCIAD3167-1GEA
57.2
EUR
TCIAD3167-1G
TCIAD3167-5G
1,6-Dibromopyrene ≥98.0% (by GC)
1,6-Dibromopyrene
Formule:
C₁₆H₈Br₂ |
MDL Nummer:
MFCD09263746 CAS nummer: 27973-29-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 228.0 to 232.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...