2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid
Leverancier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
BES, 2-(Bis(2-hydroxyethyl)amino)ethanesulphonic acid, N,N-Bis-(2-hydroxyethyl)-2-ethanesulphonic acid
9134.1
9134.2
9134.3
ROTH9134.1EA
0
EUR
ROTH9134.1
ROTH9134.2
ROTH9134.3
2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid
2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid
Formule:
C₆H₁₅NO₅S Kookpunt: 405 °C (1013 hPa) Smeltpunt: 152…156 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00007533 CAS nummer: 10191-18-1 EINECS: 233-465-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...