Dicobalt octacarbonyl stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar


Stabilisatie: Stabilised with hexane 1-5%

D3213-5G D3213-25G
TCIAD3213-5GEA 139 EUR
TCIAD3213-5G TCIAD3213-25G
Dicobalt octacarbonyl stabilised
Dicobalt octacarbonyl
Formule: C₈Co₂O₈
Smeltpunt: 51°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00016024
CAS nummer: 10210-68-1
EINECS: 233-514-0
UN: 3466
ADR: 6.1,II
Merck Index: 14,03085

Order Now

Specification Test Results

Appearance Orange to Brown to Dark red powder to crystal
Cobalt 32.0 to 36.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR