3,4-Dihydro-2H-pyran-2-methanol ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2-Hydroxymethyl-3,4-dihydro-2H-pyran, DHP Linker

D3295-1G D3295-5G
TCIAD3295-1GEA 19.8 EUR
TCIAD3295-1G TCIAD3295-5G
3,4-Dihydro-2H-pyran-2-methanol ≥97.0% (by GC)
3,4-Dihydro-2H-pyran-2-methanol
Formule: C₆H₁₀O₂
Kookpunt: 93°C
Vlampunt: 85°C
MDL Nummer: MFCD00051507
CAS nummer: 3749-36-8
EINECS: 223-150-0

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR