Diethyl (2-oxo-2-phenylethyl)phosphonate ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Diethyl Phenacylphosphonate, (2-Oxo-2-phenylethyl)phosphonic Acid Diethyl Ester
D3339-1G
D3339-5G
TCIAD3339-1GEA
93.5
EUR
TCIAD3339-1G
TCIAD3339-5G
Diethyl (2-oxo-2-phenylethyl)phosphonate ≥97.0% (by GC)
Diethyl (2-oxo-2-phenylethyl)phosphonate
Formule:
C₁₂H₁₇O₄P MW: 256.24 g/mol Kookpunt: 193°C |
MDL Nummer:
MFCD00015342 CAS nummer: 3453-00-7 |
Specification Test Results
Appearance | White to Yellow to Green clear liquid |
Purity (GC) | min. 97.0 % |
Refractive index n20/D | 1.5040 to 1.5160 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...