2-(Di-tert-butylphosphino)biphenyl ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 2-(Di-tert-Butylphosphino)biphenyl, JohnPhos, 2-(Di-t-butylphosphino))-1,1'-biphenyl
D3387-1G D3387-5G
TCIAD3387-1GEA 55 EUR
TCIAD3387-1G TCIAD3387-5G
2-(Di-tert-butylphosphino)biphenyl ≥98.0% (by GC)
2-(Di-tert-butylphosphino)biphenyl
Formule: C₂₀H₂₇P
MW: 298.40 g/mol
MDL Nummer: MFCD01862440
CAS nummer: 224311-51-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2abb2c5b1bf586c13c3c66c04804542874e7e8b6&vid=2abb2c5b1bf586c13c3c66c04804542874e7e8b6&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 85.0 to 89.0 deg-C