2,2-Dimethyl-1,3-benzodioxole ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

D3421-25G D3421-5G
TCIAD3421-25GEA 266 EUR
TCIAD3421-25G TCIAD3421-5G
2,2-Dimethyl-1,3-benzodioxole ≥98.0% (by GC)
2,2-Dimethyl-1,3-benzodioxole
Formule: C₉H₁₀O₂
Kookpunt: 182°C
Smeltpunt: 3°C
MDL Nummer: MFCD08061594
CAS nummer: 14005-14-2

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR