Dimethyl octadecanedioate ≥96.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D3540-25G
D3540-5G
TCIAD3540-25GEA
482
EUR
TCIAD3540-25G
TCIAD3540-5G
Dimethyl octadecanedioate ≥96.0% (by GC)
Dimethyl octadecanedioate
Formule:
C₂₀H₃₈O₄ Kookpunt: 207°C |
MDL Nummer:
MFCD00673461 CAS nummer: 1472-93-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 58.0 to 62.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...